APO455832084 Apollo Scientific Ltd

7-Amino-2-methyl-2H-indazole

  • MDL number MFCD03787880
  • Cas Number : 90223-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73
Synonym 2-Methyl-2H-indazol-7-amine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/7-amino-2-methyl-2h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/7-amino-2-methyl-2h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart