APO455832087 Apollo Scientific Ltd

2-Chloropyrimidine-4-carboxylic acid

  • MDL number MFCD06739041
  • Cas Number : 149849-92-3
  • Molecular Weight : 158.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-4-pyrimidinecarboxylic acid
Inchi Code 1S/C5H3ClN2O2/c6-5-7-2-1-3(8-5)4(9)10/h1-2H,(H,9,10)
Inchi Key YMDSUQSBJRDYLI-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-2-chloropyrimidine, 4-Carboxy-2-chloro-1,3-diazine
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloropyrimidine-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloropyrimidine-4-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart