APO455832091 Apollo Scientific Ltd

2-Chloropyridine-3-sulphonyl chloride

  • MDL number MFCD05664916
  • Cas Number : 6684-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-43
Synonym 2-Chloro-3-(chlorosulphonyl)pyridine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloropyridine-3-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P280;P260
MSDS https://store.apolloscientific.co.uk/product/2-chloropyridine-3-sulphonyl-chloride
pictograms GHS05;GHS07
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart