APO455832099 Apollo Scientific Ltd

2-Chloro-4-hydroxypyridine

  • Molecular Weight : 129.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 169-172
Iupac Name 2-chloro-4(3H)-pyridinone
Inchi Code 1S/C5H4ClNO/c6-5-3-4(8)1-2-7-5/h1-3H,(H,7,8)
Inchi Key VBEHFOMFHUQAOW-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 169-172

Safety Information

signalword Warning
pcodes P271;P261;P280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart