APO455832100 Apollo Scientific Ltd

6-Aminoisoquinoline

  • MDL number MFCD04114862
  • Cas Number : 23687-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214-216
Synonym Isoquinolin-6-amine, 6-Amino-2-azanaphthalene
Purity 97
Flash Point >110
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-aminoisoquinoline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-aminoisoquinoline
pictograms GHS07
hstatements H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart