APO455832101 Apollo Scientific Ltd

Isoquinoline-6-boronic acid

  • MDL number MFCD10697418
  • Cas Number : 899438-92-7
  • Molecular Weight : 172.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-isoquinolinylboronic acid
Inchi Code 1S/C9H8BNO2/c12-10(13)9-2-1-8-6-11-4-3-7(8)5-9/h1-6,12-13H
Inchi Key OBUOMTRFSCUDKS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-6-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-6-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart