APO455832102 Apollo Scientific Ltd

Isoquinoline-8-boronic acid

  • MDL number MFCD11044913
  • Cas Number : 721401-43-0
  • Molecular Weight : 172.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 8-Boronoisoquinoline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/isoquinoline-8-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/isoquinoline-8-boronic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart