APO455832107 Apollo Scientific Ltd

4-Chloro-6-methylpyrimidine

  • MDL number MFCD02322991
  • Cas Number : 3435-25-4
  • Molecular Weight : 128.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-6-methyl-1,3-diazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-6-methylpyrimidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-6-methylpyrimidine
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart