APO455832116 Apollo Scientific Ltd

2,6-Dichloro-4-methylpyridine

  • MDL number MFCD09264302
  • Cas Number : 39621-00-6
  • Molecular Weight : 162.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 63-66
Iupac Name 2,6-dichloro-4-methylpyridine
Inchi Code 1S/C6H5Cl2N/c1-4-2-5(7)9-6(8)3-4/h2-3H,1H3
Inchi Key GTHRJKYVJZJPCF-UHFFFAOYSA-N

Description

Synonym 2,6-Dichloro-4-picoline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 63-66
Product Link https://store.apolloscientific.co.uk/product/26-dichloro-4-methylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloro-4-methylpyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart