APO455832117 Apollo Scientific Ltd

4-Bromo-2-methylbenzaldehyde

  • MDL number MFCD07787171
  • Cas Number : 24078-12-4
  • Molecular Weight : 199.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-formyltoluene, 4-Bromo-o-tolualdehyde
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-methylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-methylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart