APO455832118 Apollo Scientific Ltd

6-Chloropyridine-3,4-diamine

  • MDL number MFCD04037269
  • Cas Number : 89182-17-2
  • Molecular Weight : 143.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-4,5-diaminopyridine, 6-Chloro-3,4-diaminopyridine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloropyridine-34-diamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloropyridine-34-diamine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart