APO455832125 Apollo Scientific Ltd

1H-Indazole-4-boronic acid

  • MDL number MFCD09878901
  • Cas Number : 1023595-17-6
  • Molecular Weight : 161.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 166-168
Iupac Name 1H-indazol-4-ylboronic acid
Inchi Code 1S/C7H7BN2O2/c11-8(12)6-2-1-3-7-5(6)4-9-10-7/h1-4,11-12H,(H,9,10)
Inchi Key BGZZJZIZRARGGZ-UHFFFAOYSA-N

Description

Synonym 4-Borono-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 166-168
Product Link https://store.apolloscientific.co.uk/product/1h-indazole-4-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-indazole-4-boronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart