APO455832138 Apollo Scientific Ltd

5-Chloro-2-methoxyphenol

  • MDL number MFCD11100133
  • Cas Number : 3743-23-5
  • Molecular Weight : 158.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-methoxyphenol
Inchi Code 1S/C7H7ClO2/c1-10-7-3-2-5(8)4-6(7)9/h2-4,9H,1H3
Inchi Key HIWHLFQMDDWLRB-UHFFFAOYSA-N

Description

Synonym 4-Chloro-2-hydroxyanisole, 4-Chloro-2-hydroxyphenyl methyl ether
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-methoxyphenol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-methoxyphenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart