APO455832140 Apollo Scientific Ltd

2-(Aminomethyl)-5-bromo-4-methylpyridine hydrochloride

  • MDL number MFCD17214201
  • Cas Number : 1257535-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (5-Bromo-4-methylpyridin-2-yl)methylamine hydrochloride, 5-Bromo-4-methylpicolinylamine hydrochloride
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48188

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48188
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart