APO455832141 Apollo Scientific Ltd

2-(Aminomethyl)-3-bromo-5-methylpyridine hydrochloride

  • MDL number MFCD17214207
  • Cas Number : 1257535-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Bromo-5-methylpyridin-2-yl)methylamine hydrochloride
Storage temp. Store at 2-8°C
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-aminomethyl-3-bromo-5-methylpyridine-hydrochloride

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-aminomethyl-3-bromo-5-methylpyridine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart