APO455832146 Apollo Scientific Ltd

5-Chloro-2-methoxynicotinonitrile

  • MDL number MFCD17214212
  • Cas Number : 1256812-94-8
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Chloro-2-methoxypyridine-3-carbonitrile, 5-Chloro-3-cyano-2-methoxypyridine
Storage temp. Store at 2-8
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-methoxynicotinonitrile

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-methoxynicotinonitrile
pictograms GHS07
hstatements H302;H312;H315;H319;H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart