APO455832148 Apollo Scientific Ltd

Methyl 6-chloro-3-methylpyridine-2-carboxylate

  • MDL number MFCD12756112
  • Cas Number : 878207-92-2
  • Molecular Weight : 185.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 6-chloro-3-methyl-2-pyridinecarboxylate
Inchi Code 1S/C8H8ClNO2/c1-5-3-4-6(9)10-7(5)8(11)12-2/h3-4H,1-2H3
Inchi Key XWPSWOKQWBNRDW-UHFFFAOYSA-N

Description

Synonym Methyl 6-chloro-3-methylpicolinate, 6-Chloro-2-(methoxycarbonyl)-3-methylpyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-6-chloro-3-methylpyridine-2-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-6-chloro-3-methylpyridine-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart