APO455832149 Apollo Scientific Ltd

5-Bromo-1H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD11518975
  • Cas Number : 929617-35-6
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Aza-5-bromo-1H-indazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-1h-pyrazolo34-cpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-1h-pyrazolo34-cpyridine
pictograms GHS07
hstatements H312;H315;H319;H332
Availability details for
Quantity
  • Availability
  • Add to Cart