APO455832151 Apollo Scientific Ltd

9-Bromo-10-phenylanthracene

  • MDL number MFCD00230983
  • Cas Number : 23674-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48316

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=48316
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart