APO455832153 Apollo Scientific Ltd

4-Acetyltetrahydro-2H-pyran

  • MDL number MFCD08704647
  • Cas Number : 137052-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Tetrahydro-2H-pyran-4-yl)ethan-1-one, 4-Acetyloxane
Purity 97
Density 1.024
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 90-94/15mm
Product Link https://store.apolloscientific.co.uk/product/4-acetyltetrahydro-2h-pyran

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-acetyltetrahydro-2h-pyran
pictograms GHS05;GHS07
hstatements H302+H332;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart