APO455832158 Apollo Scientific Ltd

7-Bromo-8-methylquinoline

  • MDL number MFCD09907849
  • Cas Number : 809248-61-1
  • Molecular Weight : 222.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-8-methylquinoline
Inchi Code 1S/C10H8BrN/c1-7-9(11)5-4-8-3-2-6-12-10(7)8/h2-6H,1H3
Inchi Key MLTSRPXEANUCKX-UHFFFAOYSA-N

Description

Synonym 7-Bromo-8-methyl-1-azanaphthalene
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromo-8-methylquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/7-bromo-8-methylquinoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart