APO455832167 Apollo Scientific Ltd

4-Bromo-2-chloro-6-nitroaniline

  • MDL number MFCD03426655
  • Cas Number : 34033-41-5
  • Molecular Weight : 251.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-chloro-6-nitroaniline
Inchi Code 1S/C6H4BrClN2O2/c7-3-1-4(8)6(9)5(2-3)10(11)12/h1-2H,9H2
Inchi Key IZJHDXGYFMAJQJ-UHFFFAOYSA-N

Description

Synonym 2-Amino-5-bromo-3-chloronitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-6-nitroaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-6-nitroaniline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart