APO455832172 Apollo Scientific Ltd

3-Bromo-2-hydrazino-5-methylpyridine

  • MDL number MFCD10000011
  • Cas Number : 1289007-61-9
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Bromo-5-methylpyridin-2-yl)hydrazine, 5-Bromo-6-hydrazino-3-picoline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-hydrazino-5-methylpyridine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-hydrazino-5-methylpyridine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart