APO455832176 Apollo Scientific Ltd

2-Chloro-4-hydrazinopyridine

  • MDL number MFCD08460247
  • Cas Number : 700811-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Synonym (2-Chloropyridin-4-yl)hydrazine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-hydrazinopyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-hydrazinopyridine
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart