APO455832186 Apollo Scientific Ltd

2-(3-Methyl-1,2,4-oxadiazol-5-yl)benzyl bromide

  • MDL number MFCD09064955
  • Cas Number : 892501-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-67
Synonym 5-[2-(Bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Purity 97
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=36319

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=36319
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart