APO455832187 Apollo Scientific Ltd

(4-Bromophenyl)methanesulphonyl chloride

  • MDL number MFCD04973454
  • Cas Number : 53531-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-121
Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromophenylmethanesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromophenylmethanesulphonyl-chloride
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart