APO455832188 Apollo Scientific Ltd

(3-Bromophenyl)methanesulphonyl chloride

  • MDL number MFCD06798043
  • Cas Number : 58236-74-1
  • Molecular Weight : 269.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 88-89
Iupac Name (3-bromophenyl)methanesulfonyl chloride
Inchi Code 1S/C7H6BrClO2S/c8-7-3-1-2-6(4-7)5-12(9,10)11/h1-4H,5H2
Inchi Key VCGKEAIINMDLMC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 88-89
Product Link https://store.apolloscientific.co.uk/product/3-bromophenylmethanesulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromophenylmethanesulphonyl-chloride
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart