APO455832192 Apollo Scientific Ltd

5-Amino-1-chloroisoquinoline

  • MDL number MFCD09258640
  • Cas Number : 374554-54-8
  • Molecular Weight : 178.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 158-162
Iupac Name 1-chloro-5-isoquinolinamine
Inchi Code 1S/C9H7ClN2/c10-9-7-2-1-3-8(11)6(7)4-5-12-9/h1-5H,11H2
Inchi Key DDELEGMETMZLAF-UHFFFAOYSA-N

Description

Synonym 1-Chloroisoquinolin-5-amine
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 158-162
Product Link https://store.apolloscientific.co.uk/product/5-amino-1-chloroisoquinoline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-1-chloroisoquinoline
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart