APO455832193 Apollo Scientific Ltd

3-Chloroisoquinoline

  • MDL number MFCD05982009
  • Cas Number : 19493-45-9
  • Molecular Weight : 163.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 42-45
Iupac Name 3-chloroisoquinoline
Inchi Code 1S/C9H6ClN/c10-9-5-7-3-1-2-4-8(7)6-11-9/h1-6H
Inchi Key CPCMFADZMOYDSZ-UHFFFAOYSA-N

Description

Synonym 3-Chloro-2-azanaphthalene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42-45
Product Link https://store.apolloscientific.co.uk/product/3-chloroisoquinoline

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloroisoquinoline
pictograms GHS07;GHS05
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart