APO455832194 Apollo Scientific Ltd

6-Hydroxyisoquinoline

  • MDL number MFCD04114860
  • Cas Number : 7651-82-3
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-isoquinolinol
Inchi Code 1S/C9H7NO/c11-9-2-1-8-6-10-4-3-7(8)5-9/h1-6,11H
Inchi Key GPVPDRHTRGTSIH-UHFFFAOYSA-N

Description

Synonym Isoquinolin-6-ol, 6-Hydroxy-2-azanaphthalene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-hydroxyisoquinoline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-hydroxyisoquinoline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart