APO455832195 Apollo Scientific Ltd

6-Methoxyisatoic anhydride

  • MDL number MFCD11046419
  • Cas Number : 67765-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258-264
Synonym 5-Methoxy-2H-3,1-benzoxazine-2,4(1H)-dione, 5-Methoxy-1H-benzo[d][1,3]oxazine-2,4-dione
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-methoxyisatoic-anhydride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-methoxyisatoic-anhydride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart