APO455832201 Apollo Scientific Ltd

2-(Aminomethyl)-5-bromo-3-methylpyridine hydrochloride

  • MDL number MFCD17214203
  • Cas Number : 1257535-42-4
  • Molecular Weight : 237.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-3-methyl-2-pyridinyl)methanamine hydrochloride
Inchi Code 1S/C7H9BrN2.ClH/c1-5-2-6(8)4-10-7(5)3-9;/h2,4H,3,9H2,1H3;1H
Inchi Key QNMYNZFJYJEBGB-UHFFFAOYSA-N

Description

Synonym (5-Bromo-3-methylpyridin-2-yl)methylamine hydrochloride, 5-Bromo-3-methylpicolinylamine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminomethyl-5-bromo-3-methylpyridine-hydrochloride

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminomethyl-5-bromo-3-methylpyridine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart