APO455832202 Apollo Scientific Ltd

6-Chloro-4-methylnicotinonitrile

  • MDL number MFCD17214204
  • Cas Number : 66909-35-1
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4-methylnicotinonitrile
Inchi Code 1S/C7H5ClN2/c1-5-2-7(8)10-4-6(5)3-9/h2,4H,1H3
Inchi Key DWLWCFHFMYBZKE-UHFFFAOYSA-N

Description

Synonym 6-Chloro-4-methylpyridine-3-carbonitrile, 2-Chloro-5-cyano-4-methylpyridine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chloro-4-methylnicotinonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-4-methylnicotinonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart