APO455832206 Apollo Scientific Ltd

4-(Methylsulphonyl)benzene-1,2-diamine

  • MDL number MFCD00720917
  • Cas Number : 21731-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Diamino-4-(methylsulphonyl)benzene, 3,4-Diaminophenyl methyl sulphone, 4-(Methylsulphonyl)phenylene-1,2-diamine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonylbenzene-12-diamine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonylbenzene-12-diamine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart