APO455832212 Apollo Scientific Ltd

3-(Carboxymethyl)benzeneboronic acid

  • MDL number MFCD08704669
  • Cas Number : 914397-60-7
  • Molecular Weight : 179.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-190
Synonym 2-(3-Boronophenyl)acetic acid
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-carboxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-carboxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart