APO455832219 Apollo Scientific Ltd

Methyl trans-4-aminocyclohexane-1-carboxylate hydrochloride

  • MDL number MFCD08274538
  • Cas Number : 61367-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-140
Synonym trans-1-Amino-4-(methoxycarbonyl)cyclohexane hydrochloride
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=53342

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=53342
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart