APO455832221 Apollo Scientific Ltd

3,4-Dichlorophenacyl chloride

  • MDL number MFCD03970382
  • Cas Number : 42981-08-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-47
Synonym 2-Chloro-1-(3,4-dichlorophenyl)ethan-1-one, 2,3',4'-Trichloroacetophenone
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 171-173/15mm
Product Link https://store.apolloscientific.co.uk/product/34-dichlorophenacyl-chloride

Safety Information

signalword Danger
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/34-dichlorophenacyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart