APO455832224 Apollo Scientific Ltd

2-Chloro-3-methoxypyrazine

  • MDL number MFCD02684324
  • Cas Number : 40155-28-0
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 31-32
Iupac Name 3-chloro-2-pyrazinyl methyl ether
Inchi Code 1S/C5H5ClN2O/c1-9-5-4(6)7-2-3-8-5/h2-3H,1H3
Inchi Key PUMFABSDZOUISK-UHFFFAOYSA-N

Description

Synonym 2-Chloro-3-methoxy-1,4-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 31-32
Product Link https://store.apolloscientific.co.uk/product/2-chloro-3-methoxypyrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-3-methoxypyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart