APO455832227 Apollo Scientific Ltd

2-Bromo-5-methoxypyrazine

  • MDL number MFCD08235189
  • Cas Number : 143250-10-6
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-methoxypyrazine
Inchi Code 1S/C5H5BrN2O/c1-9-5-3-7-4(6)2-8-5/h2-3H,1H3
Inchi Key YMDHKDFBWIRZAZ-UHFFFAOYSA-N

Description

Synonym 2-Bromo-5-methoxy-1,4-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-methoxypyrazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-methoxypyrazine
pictograms GHS07
hstatements H315;H319;H335;H412
Availability details for
Quantity
  • Availability
  • Add to Cart