APO455832231 Apollo Scientific Ltd

5-Bromo-3-methylpyridine-2-carbonitrile

  • MDL number MFCD11975788
  • Cas Number : 156072-86-5
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-methyl-2-pyridinecarbonitrile
Inchi Code 1S/C7H5BrN2/c1-5-2-6(8)4-10-7(5)3-9/h2,4H,1H3
Inchi Key YPCLLVYLWXIIBW-UHFFFAOYSA-N

Description

Synonym 5-Bromo-3-methylpicolinonitrile
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-methylpyridine-2-carbonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-methylpyridine-2-carbonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart