APO455832235 Apollo Scientific Ltd

Methyl 3-chloro-5-nitrobenzoate

  • MDL number MFCD01529623
  • Cas Number : 36138-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 314
Product Link https://store.apolloscientific.co.uk/product/methyl-3-chloro-5-nitrobenzoate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-3-chloro-5-nitrobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart