APO455832252 Apollo Scientific Ltd

2-Amino-6-methylpyrazine

  • MDL number MFCD08235193
  • Cas Number : 5521-56-2
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-128
Synonym 6-Methylpyrazin-2-amine, 2-Amino-6-methyl-1,4-diazine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-methylpyrazine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-methylpyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart