APO455832255 Apollo Scientific Ltd

5-Bromo-2-nitroaniline

  • MDL number MFCD09056836
  • Cas Number : 5228-61-5
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 151-153
Iupac Name 5-bromo-2-nitroaniline
Inchi Code 1S/C6H5BrN2O2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H,8H2
Inchi Key RMIFLIVHJLREFJ-UHFFFAOYSA-N

Description

Synonym 2-Amino-4-bromonitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 151-153
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-nitroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-nitroaniline
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart