APO455832265 Apollo Scientific Ltd

4-Amino-1,3-dimethyl-1H-pyrazole hydrochloride

  • MDL number MFCD02253769
  • Cas Number : 1147222-02-3
  • Molecular Weight : 147.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dimethyl-1H-pyrazol-4-ylamine hydrochloride
Inchi Code 1S/C5H9N3.ClH/c1-4-5(6)3-8(2)7-4;/h3H,6H2,1-2H3;1H
Inchi Key ZDOGXPGAEYKUJK-UHFFFAOYSA-N

Description

Synonym 1,3-Dimethyl-1H-pyrazol-4-amine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-13-dimethyl-1h-pyrazole-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-13-dimethyl-1h-pyrazole-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart