APO455832266 Apollo Scientific Ltd

4-Amino-1-ethyl-5-methyl-1H-pyrazole hydrochloride

  • MDL number MFCD04968765
  • Cas Number : 1185300-50-8
  • Molecular Weight : 161.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Ethyl-5-methyl-1H-pyrazol-4-amine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-1-ethyl-5-methyl-1h-pyrazole-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-1-ethyl-5-methyl-1h-pyrazole-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart