APO455832275 Apollo Scientific Ltd

4-Chloro-7-(morpholin-4-yl)quinazoline

  • MDL number MFCD24850028
  • Cas Number : 1334602-74-2
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-chloroquinazolin-7-yl)morpholine
Inchi Code 1S/C12H12ClN3O/c13-12-10-2-1-9(7-11(10)14-8-15-12)16-3-5-17-6-4-16/h1-2,7-8H,3-6H2
Inchi Key GKFIGWQWNINBID-UHFFFAOYSA-N

Description

Synonym 4-(4-Chloroquinazolin-7-yl)morpholine, 4-Chloro-7-(morpholin-4-yl)-1,3-benzodiazine
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-chloro-7-morpholin-4-ylquinazoline

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-7-morpholin-4-ylquinazoline
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart