APO455832277 Apollo Scientific Ltd

2-Amino-6-bromoimidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD22205889
  • Cas Number : 1392102-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Bromoimidazo[1,2-a]pyridin-2-amine hydrochloride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=50566

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=50566
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart