APO455832291 Apollo Scientific Ltd

3-Bromo-4-iodoanisole

  • MDL number MFCD12547848
  • Cas Number : 466639-53-2
  • Molecular Weight : 312.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-iodo-4-methoxybenzene
Inchi Code 1S/C7H6BrIO/c1-10-5-2-3-7(9)6(8)4-5/h2-4H,1H3
Inchi Key DIQUDEJWQWDXHP-UHFFFAOYSA-N

Description

Synonym 2-Bromo-1-iodo-4-methoxybenzene, 3-Bromo-4-iodophenyl methyl ether
Purity 97%
Flash Point 90/2mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-iodoanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-iodoanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart