APO455832305 Apollo Scientific Ltd

6-Bromo-2,7-dimethyl-3H-imidazo[4,5-b]pyridine

  • MDL number MFCD08062977
  • Cas Number : 954238-09-6
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-2,7-dimethyl-3H-imidazo[4,5-b]pyridine
Inchi Code 1S/C8H8BrN3/c1-4-6(9)3-10-8-7(4)11-5(2)12-8/h3H,1-2H3,(H,10,11,12)
Inchi Key LWFMUKDBYLABGX-UHFFFAOYSA-N

Description

Synonym 5-Bromo-2,4-dimethyl-7-azabenzimidazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-27-dimethyl-3h-imidazo45-bpyridine

Safety Information

signalword Warning
pcodes P280;P260
MSDS https://store.apolloscientific.co.uk/product/6-bromo-27-dimethyl-3h-imidazo45-bpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart