APO455832306 Apollo Scientific Ltd

2-Iodostyrene

  • MDL number MFCD00080535
  • Cas Number : 4840-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Ethenyl-2-iodobenzene, 1-Iodo-2-vinylbenzene
Density 1.6734
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 69-70/0.3mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61627

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61627
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart